Please login to your online account to display your discounted pricing

Sodium meta-Periodate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$507.79 - $1,446.61

Chemical Identifiers

Chemical Name or Material Sodium meta-Periodate
CAS 7790-28-5
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula NaIO4
Formula Weight 213.89
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004171

 
avantor performance
375604
Poly Bottle 125g Each for $507.79

02004172

 
avantor performance
375601
Poly Bottle 500g Each for $1,446.61
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 213.89
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material Sodium meta-Periodate
CAS 7790-28-5
Molecular Formula NaIO4
Packaging Poly Bottle
Quantity 125g
Specific Gravity 1L = 3.87kg