Please login to your online account to display your discounted pricing

Sodium meta-Bisulfite, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  355201

 View more versions of this product

Catalog No. 02004166

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 190.11
Chemical Name or Material Sodium meta-Bisulfite
CAS 7681-57-4
Molecular Formula Na2S2O5
Physical Form Granular
Quantity 500g
Specific Gravity 1L = 1.48kg
Packaging Poly Bottle

Specifications

Chemical Name or Material Sodium meta-Bisulfite
CAS 7681-57-4
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2S2O5
Formula Weight 190.11