Learn More

Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical™
Manufacturer: Fisher Chemical S529500
Specifications
Sodium Lauryl Sulfate | |
Sodium alkyl sulfate content: 85.0% min. (NF) | |
White | |
CH3(CH2)10CH2OSO3Na | |
MFCD00036175 | |
1325 | |
Dodecylsulfuric Acid Sodium Salt, Duponol™ C Dry, SDS, Sodium Dodecyl Sulfate | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
Pass Test (FCC) | |
3423265 | |
Solid | |
FCC/NF | |
Meets Requirements (NF) | |
59.0% min. (FCC) |
151-21-3 | |
Pass Test (FCC) | |
C12H25NaO4S | |
206°C | |
8.5 to 10 | |
500g | |
100°C | |
0.002% max. (NF) | |
sodium dodecyl sulfate | |
288.38 | |
CHEBI:8984 | |
5mg/kg max. (FCC) | |
Glass Bottle | |
8.0% max. (FCC) | |
4.0% max. (FCC) |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
sodium dodecyl sulfate |
C12H25NaO4S | |
MFCD00036175 | |
Dodecylsulfuric Acid Sodium Salt, Duponol™ C Dry, SDS, Sodium Dodecyl Sulfate | |
CHEBI:8984 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Safety and Handling

DANGER!
Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 4.1, : Flammable Solid