missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Lactate (Syrup, 60% w/w), Fisher Chemical

Manufacturer:  Fisher Chemical S326500

Catalog No. S326-500




Description

Description

Specifications

Specifications

Sodium Lactate
72-17-3,7732-18-5
CH3CH(OH)COONa
lacolin, lactic acid sodium salt, lactic acid, monosodium salt, monosodium 2-hydroxypropanoate, monosodium lactate, per-glycerin, propanoic acid, 2-hydroxy-, monosodium salt, sodium alpha-hydroxypropionate, sodium dl-lactate, sodium lactate
CC(C(=O)[O-])O.[Na+]
112.06
CHEBI:75228
Viscous Liquid
HDPE Bottle
140°C
17.2°C
500mL
Syrup, 60% (w/w)
C3H5NaO3
MFCD00065400
NGSFWBMYFKHRBD-UHFFFAOYSA-M
sodium;2-hydroxypropanoate
23666456
112.06
Pass Test
14mmHg at 20°C
Yellow
7

Chemical Identifiers

72-17-3
112.06
NGSFWBMYFKHRBD-UHFFFAOYSA-M
23666456
sodium;2-hydroxypropanoate
C3H5NaO3
MFCD00065400
lacolin, lactic acid sodium salt, lactic acid, monosodium salt, monosodium 2-hydroxypropanoate, monosodium lactate, per-glycerin, propanoic acid, 2-hydroxy-, monosodium salt, sodium alpha-hydroxypropionate, sodium dl-lactate, sodium lactate
CHEBI:75228
CC(C(=O)[O-])O.[Na+]
Safety and Handling

Safety and Handling

Recommended Storage : Refrigerate

SDS Documents