missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium L-(+)-Tartrate Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Manufacturer: Fisher Chemical S433500
Specifications
Sodium L-(+)-Tartrate Dihydrate | |
1.8g/mL | |
White | |
14 to 17% | |
≥99.0 % | |
MFCD00150035 | |
500g | |
YAPIZISBZUXUIH-DGFHWNFOSA-N | |
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate | |
131855972 | |
Powder/Solid | |
Certified |
6106-24-7 | |
7 to 9 | |
150°C | |
Pass Test | |
C4H10Na2O8 | |
Poly Bottle | |
butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, dia7c37aow, disodium +-tartrate dihydrate, disodium l-+-tartrate dihydrate, disodium tartarate dihydrate, disodium tartrate dihydrate, sodium l-+-tartrate dihydrate, sodium tartrate dihydrate mi, sodium tartrate nf, unii-dia7c37aow | |
C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na] | |
232.096 | |
230.08 | |
≥99.0% |
Chemical Identifiers
6106-24-7 | |
232.096 | |
YAPIZISBZUXUIH-DGFHWNFOSA-N | |
131855972 | |
C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na] |
C4H10Na2O8 | |
MFCD00150035 | |
butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, dia7c37aow, disodium +-tartrate dihydrate, disodium l-+-tartrate dihydrate, disodium tartarate dihydrate, disodium tartrate dihydrate, sodium l-+-tartrate dihydrate, sodium tartrate dihydrate mi, sodium tartrate nf, unii-dia7c37aow | |
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate |
Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!