missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium isopropoxide isopropanol adduct, 95%

Manufacturer:  Alfa Aesar™ 4564806

 View more versions of this product

Catalog No. AA4564806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium isopropoxide isopropanol adduct
Cream
5g
NaOCH(CH3)2·C3H7OH
UN3206
Powder
Air & Moisture Sensitive
150°C (decomposition)
NaOC3H7·C3H8O
MFCD12547728
142.17
95%

Chemical Identifiers

NaOC3H7·C3H8O
MFCD12547728
Safety and Handling

Safety and Handling

GHS H Statement
H228-H251-H314-H318
Flammable solid.
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

Documents