Please login to your online account to display your discounted pricing

Sodium isopropoxide, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sodium isopropoxide
CAS 683-60-3
Molecular Formula C3H7NaO
Linear Formula NaOCH(CH3)2
Formula Weight 82.08
MDL Number MFCD00061566
UN Number UN3206
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA8944622

  SDS
alfa aesar
8944622
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 70° to 75°C
Formula Weight 82.08
Quantity 100g
DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
Chemical Name or Material Sodium isopropoxide
UN Number UN3206
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Color White
Physical Form Crystalline
Molecular Formula C3H7NaO
MDL Number MFCD00061566
Sensitivity Air and Moisture Sensitive
Health Hazard 1 Danger
Linear Formula NaOCH(CH3)2
TSCA TSCA
CAS 683-60-3
Health Hazard 2 GHS H Statement
H251-H228-H314-H318;
Self-heating: may catch fire.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

GHS H Statement
H251-H228-H314-H318;
Self-heating: may catch fire.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.