missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium iodide, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1360406

 View more versions of this product

Catalog No. AA1360406



Description

Description

  • Ampouled under argon
Specifications

Specifications

Sodium iodide
7681-82-5
INa
14,8631
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium;iodide
5238
149.89
99.999%
−10 Mesh
Ampule
Light Sensitive & Hygroscopic
White
5g
Ultra dry
(metals basis)
MFCD00003532
sodium iodide, ioduril, sodium iodide, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
[Na+].[I-]
149.894
CHEBI:33167
Beads
3.67g/mL
Odorless
1.7745
1300°C to 1304°C
651°C

Chemical Identifiers

7681-82-5
149.894
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
5238
sodium;iodide
INa
MFCD00003532
sodium iodide, ioduril, sodium iodide, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
CHEBI:33167
[Na+].[I-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WB6475000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents