Learn More
Alfa Aesar™ Sodium iodide, ultra dry, 99.999% (metals basis)
Manufacturer: Alfa Aesar™ 1360422
Description
- Ampouled under argon
Specifications
Sodium iodide | |
7681-82-5 | |
INa | |
14,8631 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
sodium;iodide | |
5238 | |
149.89 | |
99.999% | |
−10 Mesh | |
Ampule | |
Light Sensitive & Hygroscopic | |
White | |
100g |
Ultra dry | |
(metals basis) | |
MFCD00003532 | |
sodium iodide, ioduril, sodium iodide, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
[Na+].[I-] | |
149.894 | |
CHEBI:33167 | |
Beads | |
3.67g/mL | |
Odorless | |
1.7745 | |
1300°C to 1304°C | |
651°C |
Chemical Identifiers
7681-82-5 | |
149.894 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
5238 | |
sodium;iodide |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium iodide, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
CHEBI:33167 | |
[Na+].[I-] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : WB6475000
TSCA : TSCA
storageNote1 :
- Ampouled under argon