Please login to your online account to display your discounted pricing

Sodium iodide, Ultra dry, 99.99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sodium iodide
CAS 7681-82-5
Purity Grade Ultra dry
Physical Form Powder
Assay Percent Range 99.99% (metals basis)
Molecular Formula NaI
Formula Weight 149.89
MDL Number MFCD00003532
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4476614

  SDS
alfa aesar
4476614
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4476622

  SDS
alfa aesar
4476622
Ampuled under Argon 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4476636

  SDS
alfa aesar
4476636
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium iodide
CAS 7681-82-5
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
EINECS Number 231-679-3
Purity Grade Ultra dry
Physical Form Powder
Assay Percent Range 99.99% (metals basis)
Molecular Formula NaI
Formula Weight 149.89
MDL Number MFCD00003532
Odor Odorless
Color White
Quantity 25g
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.