missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodide, Ultra dry, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4476636

 View more versions of this product

Catalog No. AA4476636



Specifications

Specifications

Sodium iodide
99.99% (metals basis)
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium;iodide
5238
149.89
Ultra dry
White
7681-82-5
INa
sodium iodide, ioduril, sodium iodide, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
[Na+].[I-]
149.894
CHEBI:33167
Powder
Odorless
500g

Chemical Identifiers

7681-82-5
149.894
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
5238
sodium;iodide
INa
MFCD00003532
sodium iodide, ioduril, sodium iodide, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
CHEBI:33167
[Na+].[I-]
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 231-679-3

SDS Documents