Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium iodide, ultra dry, 99.98% (metals basis)

Chemical Identifiers

Chemical Name or Material Sodium iodide
CAS 7681-82-5
Name Note Ultra dry
Formula Weight 149.89
MDL Number MFCD00003532
Merck Index 14,8631
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4344714

  SDS
alfa aesar™
4344714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 149.89
Boiling Point 1300° to 1304°C
Packaging Ampule
TSCA TSCA
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WB6475000
Chemical Name or Material Sodium iodide
Mesh Size −10 Mesh
Odor Odorless
Name Note Ultra dry
Physical Form Beads
Refractive Index 1.7745
Assay 99.98%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00003532
CAS 7681-82-5
Merck Index 14,8631
Melting Point 651°C
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1
  • Ampouled under argon
Density 3.67g/mL

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon