Please login to your online account to display your discounted pricing

Sodium Iodide AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Iodide
CAS 7681-82-5
Physical Form Granular
Molecular Formula NaI
Formula Weight 149.89
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK1141212

 
avantor performance
114106
Glass 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium Iodide
CAS 7681-82-5
Physical Form Granular
Molecular Formula NaI
Formula Weight 149.89
Packaging Glass
Quantity 2.5kg