missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Iodide, ACS Grade, ≥ 99.5%, LabChem™

$410.14 - $410.14

Chemical Identifiers

CAS 7681-82-5
CAS Max % 100
Linear Formula NaI
Formula Weight 149.89
Chemical Name or Material Sodium Iodide
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC246451 Lab Chem
LC246451
500g Each for $410.14
Specifications Description & Specifications

Chemical Identifiers

7681-82-5
NaI
Sodium Iodide
100
149.89
ACS Grade

Specifications

7681-82-5
Crystals
149.89
Passes Test
Soluble in water
Room Temperature; Protect from light
1,304°C
651°C
Sodium Iodide
ACS Grade
100
NaI
3.667g/cm3
Amber Glass
3.667g/cm3
Iodine vapor
White
500g
≥99.5%

Safety and Handling

GHS H Statement
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Collect spillage.

Warning

EINECSNumber : 231-668-3