missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodide, 99+%, pure, anhydrous, ACROS Organics™

Manufacturer:  Acros Organics 203180010

 View more versions of this product

Catalog No. AC203180010



Specifications

Specifications

Sodium iodide
7681-82-5
INa
MFCD00003532
15,8765
Solubility in water: 184g/100mL (25°C). Other solubilities: soluble in alcohol,glycerol and acetone
[Na+].[I-]
149.89
CHEBI:33167
Crystalline Powder
Pure
3% max. (1 g, 105°C, 3 hrs)
Solubility in water: 184 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and acetone
1300.0°C
661.0°C
Anhydrous
99+%
NaI
01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86
iodure de sodium
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium;iodide
5238
149.89
≥99%
3.6600g/mL
Glass bottle
3.66
White
1kg

Chemical Identifiers

7681-82-5
149.89
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
5238
sodium;iodide
INa
MFCD00003532
iodure de sodium
CHEBI:33167
[Na+].[I-]
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents