missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodide, 99.9% (metals basis), Alfa Aesar™

$96.85 - $1,209.89

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym iodure de sodium, ioduril, jodid sodny, jodid sodny czech, natrii iodidum, sodium iodide, sodium monoiodide, sodiumiodide, soiodin
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1166522
View Documents
Thermo Fisher Scientific
1166522
100g Each for $96.85
 
AA1166536
View Documents
Thermo Fisher Scientific
1166536
500g Each for $375.93
 
AA11665A3
View Documents
Thermo Fisher Scientific
11665A3
2kg Each for $1,209.89
 
Specifications

Chemical Identifiers

7681-82-5
149.894
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
5238
sodium;iodide
INa
MFCD00003532
iodure de sodium, ioduril, jodid sodny, jodid sodny czech, natrii iodidum, sodium iodide, sodium monoiodide, sodiumiodide, soiodin
CHEBI:33167
[Na+].[I-]

Specifications

7681-82-5
INa
White powder or beads
Very soluble in water and liquid ammonia. Freely soluble in acetone,formic acid,formamide,methanol,and acetonitrile. Soluble in N,N-dimethylformamide.
[Na+].[I-]
149.894
CHEBI:33167
Powder
Odorless
Hygroscopic
White
100g
99.9% (metals basis)
MFCD00003532
iodure de sodium, ioduril, jodid sodny, jodid sodny czech, natrii iodidum, sodium iodide, sodium monoiodide, sodiumiodide, soiodin
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium;iodide
5238
149.89
3.67
1.7745
1300°C to 1304°C
651°C
Sodium iodide
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-679-3

RTECSNumber : WB6475000

TSCA : Yes

Recommended Storage : Ambient temperatures