Please login to your online account to display your discounted pricing

Sodium ICP Standard, 1mL = 1mg Na (1,000ppm Na), Ricca Chemical

$167.09 - $311.39

Chemical Identifiers

Chemical Name or Material Sodium ICP Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.59
CAS Max % 4.77
CAS 497-19-8
CAS Min % 0.21
CAS Max % 0.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PNA1KN-100

 
ricca chemical company
RIPNA1KN100
Poly natural 100mL Each for $167.09

PNA1KN-500

 
ricca chemical company
RIPNA1KN500
Poly natural 500mL Each for $311.39
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Concentration or Composition Notes 1,000ppm Na
Equivalence 1 mL = 1 mg Na
CAS 7732-18-5
CAS Min % 93.0
CAS 497-19-8
CAS Max % 97.0
CAS Max % 4.77
CAS Min % 0.21
CAS Min % 4.59
CAS Max % 0.22
Packaging Poly natural
Chemical Name or Material Sodium ICP Standard
CAS 7697-37-2