missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium ICP Standard, 10,000 ppm Na in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PNA10KN50
Specifications
Sodium ICP Standard | |
Natural Poly Bottle | |
88.0,7.62,2.04 | |
Colorless | |
CNa2O3 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
1 mL = 10 mg Na |
497-19-8,7697-37-2,7732-18-5 | |
50 mL | |
92.0,7.93,2.12 | |
10,000ppm Na | |
MFCD00003494 | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
Liquid |
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
CHEBI:29377 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
10340 | |
disodium carbonate |