Learn More
Sodium hypochlorite, 11-15% available chlorine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03336936
Description
Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hypochlorite | |
7681-52-9 | |
Liquid | |
ClNaO | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
sodium;hypochlorite | |
23665760 | |
74.44 | |
Light yellow liquid | |
1.2 g/cm3 at 20°C |
14.5% available chlorine | |
Yellow | |
500 g | |
MFCD00011120 | |
Miscible with water. | |
[O-]Cl.[Na+] | |
74.439 | |
CHEBI:32146 | |
14.5% (available chlorine) | |
23 hPa (17mm Hg) at 20°C |
Chemical Identifiers
7681-52-9 | |
MFCD00011120 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
CHEBI:32146 |
ClNaO | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
23665760 |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : DOT Class: 8; Packing Group: III
EINECSNumber : 231-668-3
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only