Please login to your online account to display your discounted pricing

Sodium Hydroxide-Thiosulfate, 40%-2.5% (w/v) Aqueous Solution, Ricca Chemical

For Kjeldahl Nitrogen Using Mercury-Based Catalysts

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide-Thiosulfate Solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7490-5

 
ricca chemical company
RI74905
Cubitainer™ 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7490-1

 
ricca chemical company
RI74901
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 20L
Chemical Name or Material Sodium Hydroxide-Thiosulfate Solution
Concentration or Composition (by Analyte or Components) 40% (w/v) Sodium Hydroxide - 2.5% (w/v) Sodium Thiosulfate
Packaging Cubitainer™