Please login to your online account to display your discounted pricing

Sodium Hydroxide on Support, Granulated (1.6 to 3mm), For Elementary Analysis, EMD Millipore™

$314.00 - $783.09

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide on Support
CAS 1310-73-2
CAS 14807-96-6
UN Number UN 1823
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1015640250

 
emd millipore corporation
1015640250
Each for $314.00

M1015641000

 
emd millipore corporation
1015641000
Each for $783.09
Description & Specifications

Specifications

DOT Information DOT Class: 8, Packaging Group: II
Physical Form Solid
Chemical Name or Material Sodium Hydroxide on Support
Health Hazard 1 H290: May be corrosive to metals.
H314: Causes severe skin burns and eye damage.
CAS 14807-96-6
CAS 1310-73-2
Particle Size 1.6 to 3mm
UN Number UN 1823
Packaging Plastic bottle

For elementary analysis

Safety and Handling

H290: May be corrosive to metals.
H314: Causes severe skin burns and eye damage.