missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide on support, Granulated (0.8 to 1.6mm), For Elementary Analysis, MilliporeSigma™

Manufacturer:  MilliporeSigma 1015671000

 View more versions of this product

Catalog No. M1015671000



Description

Description

For elementary analysis
Specifications

Specifications

Sodium Hydroxide on Support
Plastic bottle
1kg
UN 1823
[OH-].[Na+]
39.997
CHEBI:32145
1310-73-2,14807-96-6
0.8 to 1.6mm
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Solid

Chemical Identifiers

1310-73-2
39.997
14798
sodium;hydroxide
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
CHEBI:32145
[OH-].[Na+]
Safety and Handling

Safety and Handling

H290: May be corrosive to metals.
H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

SDS Documents
Promotions

Promotions