Please login to your online account to display your discounted pricing

Sodium Hydroxide StandAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
Name Note 10.0 Normal Volumetric Solution
CAS 1310-73-2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MKH3851

 
avantor performance
H38505
Poly 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MKH3854

 
avantor performance
H38507
Poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Sodium Hydroxide
Name Note 10.0 Normal Volumetric Solution
CAS 1310-73-2
Packaging Poly