Please login to your online account to display your discounted pricing

Sodium Hydroxide StandAR, Macron Fine Chemicals™


macron

Quantity 4L
Packaging Poly

Manufacturer: avantor performance  H38507

 View more versions of this product

Catalog No. MKH3854

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 4L
Chemical Name or Material Sodium Hydroxide
Name Note 10.0 Normal Volumetric Solution
CAS 1310-73-2
Packaging Poly

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note 10.0 Normal Volumetric Solution
CAS 1310-73-2