Please login to your online account to display your discounted pricing

Sodium Hydroxide Solution AR, Macron Fine Chemicals™


macron

Quantity 4L
Packaging Poly

Manufacturer: avantor performance  770508

 View more versions of this product

Catalog No. MK77054

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 4L
Chemical Name or Material Sodium Hydroxide Solution
Assay Percent Range 50%
Molecular Formula NaOH
CAS 1310-73-2
Packaging Poly

Specifications

Chemical Name or Material Sodium Hydroxide Solution
CAS 1310-73-2
Assay Percent Range 50%
Molecular Formula NaOH