Please login to your online account to display your discounted pricing

Sodium Hydroxide Solution AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide Solution
CAS 1310-73-2
Assay Percent Range 50%
Molecular Formula NaOH
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK77051

 
avantor performance
770506
Poly 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK77054

 
avantor performance
770508
Poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK7705500

 
avantor performance
770504
Poly 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Sodium Hydroxide Solution
CAS 1310-73-2
Assay Percent Range 50%
Packaging Poly
Molecular Formula NaOH