Please login to your online account to display your discounted pricing

Sodium Hydroxide Solution (50% w/w/Certified), Fisher Chemical


Quantity 20L
Packaging Poly Pail

Manufacturer: fisher scientific company  SS25420

 View more versions of this product

Catalog No. SS254-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Melting Point 5°C
Boiling Point 141.7°C
Purity Grade Certified Grade
Quantity 20L
DOT Information DOT Class 8, : Corrosive
Assay Inclusive between 50%
Physical Form Liquid
pH 12.0
Color Clear
Vapor Pressure 3mmHg at 37°C
Identification Pass Test
Potassium (K) 0.025% max.
Heavy Metals (as Ag) 0.001% max.
Ammonium Hydroxide ppt 0.010% max.
Nitrogen Compounds 5ppm max.
Additional Information Vapor Pressure: 3mmHg at 37°C
Synonym Caustic Soda Solution
Carbonate 0.10% max.
MDL Number MFCD00003548
ChemAlert Storage Symbol White
Phosphate 5ppm max.
Chloride 0.005% max.
Iron (Fe) 5ppm max.
Packaging Poly Pail
Silica 0.01% max.
Sulfate 0.002% max.
CAS 7732-18-5
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1
Name Note 50% (w/w)
Cleanup Symbol Caustic
Chemical Name or Material Sodium Hydroxide Solution
CAS 1310-73-2

Specifications

Chemical Name or Material Sodium Hydroxide Solution
Name Note 50% (w/w)
CAS 1310-73-2
CAS 7732-18-5
Purity Grade Certified Grade
Assay Inclusive between 50%
MDL Number MFCD00003548
Synonym Caustic Soda Solution
Accessories
 

Emergency Overview
Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1

DANGER!

 

SDS & Safety Documents