Please login to your online account to display your discounted pricing

Sodium Hydroxide Solution (10N/Certified), Fisher Chemical


Quantity 4L
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSS2554

 View more versions of this product

Catalog No. SS255-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Melting Point −10°C
Boiling Point >100°C
Purity Grade Certified Grade
Quantity 4L
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Density 1.32g/mL
Vapor Pressure 14mmHg
pH 12.0
Color Clear
Cleanup Symbol Caustic
Molecular Formula NaOH
MDL Number MFCD00003548
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Name Note 10N
Concentration 9.95 to 10.05
CAS 1310-73-2
ChemAlert Storage Symbol White
UN Number 1824
Synonym Caustic Soda Solution
Nitrogen Compounds 3ppm max.
CAS 7732-18-5
Chemical Name or Material Sodium Hydroxide Solution

Specifications

Chemical Name or Material Sodium Hydroxide Solution
Name Note 10N
CAS 1310-73-2
CAS 7732-18-5
Purity Grade Certified Grade
Molecular Formula NaOH
MDL Number MFCD00003548
Synonym Caustic Soda Solution
Accessories
   

SDS & Safety Documents