Learn More
Sodium Hydroxide Solution (0.25N (N/4)/Certified), Fisher Chemical™
A volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Supplier: Thermo Fisher Scientific FLSS2721
Specifications
13 | |
Colorless | |
1310-73-2,7732-18-5 | |
MFCD00003548 | |
1824 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Certified | |
Poly Bottle |
Sodium Hydroxide Solution | |
0.25N (N/4) | |
HNaO | |
0.2495 to 0.2505N | |
1.010g/cm³ | |
1 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Pass Test | |
Liquid |
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Safety and Handling
WARNING!
Emergency Overview
Causes eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:0
Instability:0
DOTInformation : DOT Class 8, : Corrosive