Please login to your online account to display your discounted pricing

Sodium Hydroxide Solution 0.1N, EMD Millipore


Quantity 4L
Packaging Poly Bottle

Manufacturer: emd millipore corporation  SX0607C3

 View more versions of this product

Catalog No. MSX0607C3

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide Solution 0.1N
Quantity 4L
Packaging Poly Bottle

Specifications

Chemical Name or Material Sodium Hydroxide Solution 0.1N