missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide Solution 0.1N, EMD Millipore

$47.34 - $85.26

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide Solution 0.1N
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MSX0607C3 Emd Millipore Corporation
SX0607C3
Poly Bottle 4L Each for $85.26
MSX0607C6 Emd Millipore Corporation
SX0607C6
Poly Bottle 1L Each for $47.34
Specifications Description & Specifications

Chemical Identifiers

Sodium Hydroxide Solution 0.1N

Specifications

4L
Poly Bottle
Sodium Hydroxide Solution 0.1N