missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide-Sodium Thiosulfate, For Nitrogen (Organic), Certified, 12.5N ±1.0N, LabChem™
Supplier: LabChem LC246005
Specifications
Sodium Hydroxide-Sodium Thiosulfate | |
For Nitrogen (Organic) | |
1310-73-2,10102-17-7 | |
12.5N ±1.0N | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
40 | |
Passes Test | |
1.39g/mL | |
15cSt | |
20 L |
Colorless | |
61.5,36,2.5 | |
NaOH | |
1.39g/mL | |
Soluble in water | |
Certified | |
Cubitainer™ | |
Traceable to NIST | |
Sodium oxide; Thermal decomposition generates: Corrosive vapors; Sulfur compounds | |
Liquid |