Please login to your online account to display your discounted pricing

Sodium Hydroxide (Pellets), ACS Reagent Grade, Ricca Chemical


Quantity 500g
Packaging Poly Wide mouth

Manufacturer: ricca chemical company  RIRDCS0530500B1

 View more versions of this product

Catalog No. RDCS0530500

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
Color White
Quantity 500g
Name Note Pellets
CAS 1310-73-2
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets or Flakes
Packaging Poly Wide mouth

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note Pellets
CAS 1310-73-2
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets or Flakes