Please login to your online account to display your discounted pricing

Sodium Hydroxide (Pellets), ACS Reagent Grade, Ricca Chemical


Quantity 2.5kg
Packaging Plastic container

Manufacturer: ricca chemical company  RIRDCS053025A5

 View more versions of this product

Catalog No. RDCS053025

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets or Flakes
Color White
Quantity 2.5kg
Name Note Pellets
CAS 1310-73-2
CAS Min % 98.0
Packaging Plastic container
CAS Max % 100.0
Chemical Name or Material Sodium Hydroxide

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note Pellets
CAS 1310-73-2
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets or Flakes