Please login to your online account to display your discounted pricing

Sodium Hydroxide (Pellets), ACS Reagent Grade, Ricca Chemical


Quantity 125g
Packaging Poly Wide mouth

Manufacturer: ricca chemical company  RIRDCS0530125B1

 View more versions of this product

Catalog No. RDCS0530125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Deliquescent Pellets or Flakes
Purity Grade ACS Reagent Grade
Color White
Quantity 125g
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Name Note Pellets
CAS Min % 98.0
CAS Max % 100.0
Packaging Poly Wide mouth

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note Pellets
CAS 1310-73-2
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets or Flakes