Please login to your online account to display your discounted pricing

Sodium Hydroxide (Pellets), ACS Reagent Grade, Ricca Chemical


Quantity 12kg
Packaging Fiberboard drum

Manufacturer: ricca chemical company  RIRDCS053012F4

 View more versions of this product

Catalog No. RDCS053012F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Deliquescent Pellets or Flakes
Purity Grade ACS Reagent Grade
Color White
Quantity 12kg
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Name Note Pellets
CAS Min % 98.0
Packaging Fiberboard drum
CAS Max % 100.0

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note Pellets
CAS 1310-73-2
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Deliquescent Pellets or Flakes