missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide NF GenAR Pellet, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 1310-73-2
Physical Form Pellet
Molecular Formula NaOH
Formula Weight 40
Chemical Name or Material Sodium Hydroxide
Grade NF
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7772212 Avantor Performance
777206
Poly Bottle 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1310-73-2
NaOH
Sodium Hydroxide
Pellet
40
NF

Specifications

2.5kg
Pellet
40
Sodium Hydroxide
1310-73-2
NaOH
Poly Bottle
NF