Please login to your online account to display your discounted pricing

Sodium Hydroxide (NF/EP/BP/FCC), Fisher Chemical


Quantity 1kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  S3991

 View more versions of this product

Catalog No. S3991

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 40
Purity Grade BP/EP/FCC/NF
Boiling Point 1390°C
Melting Point 318°C
Quantity 1kg
DOT Information DOT Class 8, : Corrosive
Assay 97.0 to 100.5% (BP)
Physical Form Solid
pH 14
Color White
Density 2.13g/cm³
Vapor Pressure 1mmHg at 739°C
Sulfate 50ppm max. (BP)
Residual Solvents Meets Requirements (BP)
Lead (Pb) 2mg/kg max. (FCC)
Insoluble Subs. and Organic Matter Pass Test (FCC)
Potassium (K) Pass Test (NF)
Identification Pass Test (BP)
Heavy Metals 0.003% max. (NF), 20ppm max. (BP/EP)
Carbonate 2.0% max. (BP)
Appearance of Solution Pass Test (BP)
Endotoxin Report EU/mg (NF)
Mercury (Hg) 0.1mg/kg max. (FCC)
CAS 497-19-8
Sodium Carbonate 3.0% max. (NF)
Heavy Metals (as Pb) 0.003% max. (NF)
Chloride 50ppm max. (BP)
Arsenic (As) 3mg/kg max. (FCC)
Cleanup Symbol Caustic
MDL Number MFCD00003548
Iron (Fe) 10ppm max. (BP)
Additional Information Vapor Pressure: 1mmHg at 739°C
Bacterial Endotoxins Report EU/mg (EP)
Packaging Poly Bottle
Molecular Formula NaOH
Health Hazard 3 Emergency Overview
Causes severe burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1
Health Hazard 2 DANGER!
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
CAS 497-19-8
Purity Grade BP/EP/FCC/NF
Assay 97.0 to 100.5% (BP)
Molecular Formula NaOH
Formula Weight 40
MDL Number MFCD00003548
Accessories

Emergency Overview
Causes severe burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1

DANGER!

 

SDS & Safety Documents