Please login to your online account to display your discounted pricing

Sodium Hydroxide, MP Biomedicals


Quantity 5g

Manufacturer: mp biomedicals  15349505

 View more versions of this product

Catalog No. ICN15349505

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 40
Quantity 5g
Recommended Storage Room Temperature
Chemical Name or Material Sodium Hydroxide
Molecular Formula NaOH
Name Note Anhydrous
CAS 1310-73-2

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note Anhydrous
CAS 1310-73-2
Molecular Formula NaOH
Formula Weight 40
Accessories

Room Temperature