Please login to your online account to display your discounted pricing

Sodium Hydroxide, in Methanol, Certified, 0.0100N ±0.0001N (0.01M) , LabChem™

$68.26 - $124.91

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 67-56-1
CAS Max % 99.96
CAS 1310-73-2
CAS Max % 0.04
Name Note in Methanol
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC245202

 
lab chem
LC245202
1L Each for $68.26

LC245204

 
lab chem
LC245204
4L Each for $124.91
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 40
Quantity 1L
Chemical Name or Material Sodium Hydroxide
Decomposition Information Carbon monoxide; Carbon dioxide
Name Note in Methanol
Concentration 0.0100N ±0.0001N (0.01M)
Physical Form Liquid
Linear Formula NaOH
CAS 67-56-1
UN Number UN1993
Identification Passes Test
Synonym Caustic Soda
Health Hazard 1 Danger
Packaging Poly Bottle
CAS Max % 99.96
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
CAS 1310-73-2
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, extinguishing powder, sand to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Storage Information Room Temperature
CAS Max % 0.04
Traceability to NIST Traceable to NIST
Color Clear, colorless
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, extinguishing powder, sand to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).