missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide (low chloride), ACS, 97.0% min, Alfa Aesar™

For Use in Chemical Manufacture, Rayon and Cellophane, As a Neutralizing Agent in Petroleum Refining

Manufacturer:  Alfa Aesar 1345536

 View more versions of this product

Catalog No. AA1345536



Specifications

Specifications

Sodium hydroxide
1310-73-2
HNaO
White pellets
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
ACS
Odorless
White
(low chloride)
97.0% min.
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Pellets
2.13g/cm3
3.5 hPa (3mm Hg) at 800°C
500g

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 215-185-5

Documents