Please login to your online account to display your discounted pricing

Sodium hydroxide, extra pure, 50 wt% solution in water, ACROS Organics™

$33.11 - $136.05

Chemical Identifiers

Chemical Name or Material Sodium hydroxide
Name Note 50 wt% solution in water
CAS 1310-73-2
CAS 7732-18-5
Purity Grade Extra Pure
Assay 49.0 to 51.0% (Acidimetry)
Molecular Formula HNaO
Linear Formula NaOH
Formula Weight 40
MDL Number MFCD00003548
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC259860251

  SDS
thermo fisher scientific
AC259860251
Plastic Jerrycan 25L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC259860010

  SDS
thermo fisher scientific
AC259860010
Plastic Bottle 1L Each for $61.10

AC259860025

  SDS
thermo fisher scientific
259860025
Plastic Bottle 2.5L Each for $136.05

AC259860250

  SDS
fisher scientific acros
259860250
Plastic Bottle 25mL Each for $33.11
Description & Specifications

Specifications

Assay 49.0 to 51.0% (Acidimetry)
Formula Weight 40
Physical Form Powder
Purity Grade Extra Pure
Quantity 25L
Color White to light gray
Merck Index 15, 8761
Packaging Plastic Jerrycan
TSCA TSCA
EINECS Number 215-185-5
RTECS Number WB4900000
Specific Gravity about 1.525 (20°C)
Density 1.5000g/mL
Fieser 01,1083; 05,616; 07,336; 08,460
UN Number 1824
Heavy Metals 10ppm max.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003548
Sodium Sulfate sodium sulfate (Na2SO4): 0.035% max.
Iron (Fe) 5ppm max.
Sodium Carbonate 0.1 % max. (Na2CO3)
Synonym Caustic soda
Sodium Chloride 100ppm (NaCl) max.
CAS 7732-18-5
Chemical Name or Material Sodium hydroxide
Name Note 50 wt% solution in water
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Linear Formula NaOH
Molecular Formula HNaO
CAS 1310-73-2
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.