Please login to your online account to display your discounted pricing

Sodium Hydroxide - EDTA Solution, 10N, Ricca Chemical

$168.76 - $168.76

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide - EDTA Solution
CAS 1310-73-2 (Sodium Hydroxide)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7478-32

 
ricca chemical company
RI747832
Poly natural 1L Each for $168.76

7478-16

 
ricca chemical company
RI747816
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 1L
Concentration or Composition Notes 10 N
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural
Chemical Name or Material Sodium Hydroxide - EDTA Solution
CAS 1310-73-2 (Sodium Hydroxide)