Please login to your online account to display your discounted pricing

Sodium Hydroxide, Dilution Solution for Cyanide, Certified, 0.16% (w/v) ±0.01%, 1.6g/L, LabChem™

$73.80 - $73.80

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 99.84
CAS 1310-73-2
CAS Max % 0.16
Name Note Dilution Solution for Cyanide
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC239502

 
lab chem
LC239502
1L Each for $73.80
Description & Specifications

Specifications

Formula Weight 40
Assay 0.16% ± 0.01% (w/v)
Purity Grade Certified Grade
Quantity 1L
CAS 1310-73-2
CAS Max % 99.84
CAS 7732-18-5
CAS Max % 0.16
Specific Gravity 1g/mL
Color Clear, colorless
Concentration 1.6g/L
Synonym Caustic Soda
Name Note Dilution Solution for Cyanide
Packaging Poly Bottle
Decomposition Information Sodium oxide
Identification Passes Test
Traceability to NIST Traceable to NIST
Solubility Soluble in water
Viscosity 1.02cSt
Density 1g/mL
Linear Formula NaOH
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Storage Information Room Temperature
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.