Please login to your online account to display your discounted pricing

Sodium Hydroxide Dilution Solution, 0.16% (w/v) Aqueous Solution, ASTM/APHA, Ricca Chemical

$48.15 - $48.15

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide Dilution Solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7474-1

 
ricca chemical company
RI74741
Plastic 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7474-16

 
ricca chemical company
RI747416
Plastic 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7474-32

 
ricca chemical company
RI747432
Plastic 1L Each for $48.15
Description & Specifications

Specifications

Chemical Name or Material Sodium Hydroxide Dilution Solution
Quantity 4L
Concentration or Composition (by Analyte or Components) 0.16% (w/v) Aqueous Solution
Packaging Plastic
Methods ASTM/APHA for Cyanide