Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 4.00N ±0.02N (4.0M), LabChem™

$81.97 - $184.43

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 86.1
CAS 1310-73-2
CAS Max % 13.9
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC244302

 
lab chem
LC244302
1L Each for $81.97

LC244304

 
lab chem
LC244304
4L Each for $184.43
Description & Specifications

Specifications

Formula Weight 40
Purity Grade Certified Grade
Quantity 1L
Color Clear, colorless
CAS 1310-73-2
Packaging Poly Bottle
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Linear Formula NaOH
Solubility Soluble in water
Concentration 4.00N ±0.02N (4.0M)
Synonym Caustic Soda
Traceability to NIST Traceable to NIST
CAS Max % 86.1
CAS 7732-18-5
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Chemical Name or Material Sodium Hydroxide
Identification Passes Test
Storage Information Room Temperature
Viscosity 2.2cSt
Decomposition Information Sodium oxide; Thermal decomposition generates: Corrosive vapors
UN Number UN1824
Density 1.15g/mL
CAS Max % 13.9
Health Hazard 1 Danger
Physical Form Liquid
Specific Gravity 1.15g/mL
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.