Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 3.00N ±0.03N (3.0M), LabChem™

$32.34 - $190.31

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 89.53
CAS 1310-73-2
CAS Max % 10.47
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC244201

 
lab chem
LC244201
Each for $32.34

LC244202

 
lab chem
LC244202
Each for $43.23

LC244204

 
lab chem
LC244204
Each for $91.00

LC244206

 
lab chem
LC244206
Each for $190.31
Description & Specifications

Specifications

Chemical Name or Material Sodium Hydroxide
Color Clear, colorless
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 89.53
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 1310-73-2
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 10.47
Purity Grade Certified Grade
Concentration 3.00N ±0.03N (3.0M)
Physical Form Liquid
Density 1.13g/mL
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Identification Passes Test
Synonym Caustic Soda
Packaging Poly Bottle
Solubility Soluble in water
Storage Information Room Temperature
Specific Gravity 1.13g/mL
Traceability to NIST Traceable to NIST
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.