Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 24% (w/v) ±1%, LabChem™

$61.94 - $61.94

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 80.27
CAS 1310-73-2
CAS Max % 19.73
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC241002

 
lab chem
LC241002
1L Each for $61.94
Description & Specifications

Specifications

Formula Weight 40
Assay 24% ± 1% (w/v)
Purity Grade Certified Grade
Quantity 1L
CAS 7732-18-5
UN Number UN1824
Packaging Poly Bottle
CAS Max % 80.27
Specific Gravity 1.21g/mL
Color Clear, colorless
Linear Formula NaOH
Synonym Caustic Soda
CAS 1310-73-2
Storage Information Room Temperature
Health Hazard 1 Danger
Identification Passes Test
Chemical Name or Material Sodium Hydroxide
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS Max % 19.73
Physical Form Liquid
Traceability to NIST Traceable to NIST
Density 1.21g/mL
Solubility Soluble in water
Viscosity 3.78cSt
Decomposition Information Sodium oxide; Thermal decomposition generates: Corrosive vapors
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.