Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 18.0% (w/w) ±0.5% (w/w), 5.39N (5.39M), LabChem™

$76.00 - $76.00

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 82
CAS 1310-73-2
CAS Max % 18
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC240852

 
lab chem
LC240852
1L Each for $76.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 18.0% ± 0.5% (w/w)
Formula Weight 40
Quantity 1L
Chemical Name or Material Sodium Hydroxide
Decomposition Information Sodium oxide; Thermal decomposition generates: Corrosive vapors
Viscosity 3.09cSt
Concentration 5.39N (5.39M)
Physical Form Liquid
Density 1.2g/mL
Linear Formula NaOH
CAS 7732-18-5
UN Number UN1824
Identification Passes Test
Synonym Caustic Soda
Health Hazard 1 Danger
Packaging Poly Bottle
CAS Max % 82
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 1310-73-2
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Storage Information Room Temperature
Specific Gravity 1.2g/mL
CAS Max % 18
Traceability to NIST Traceable to NIST
Color Clear, colorless
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.