Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 0.400N ±0.004N (0.4M), LabChem™

$46.03 - $265.57

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 98.43
CAS 1310-73-2
CAS Max % 1.57
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC243202

 
lab chem
LC243202
Each for $46.03

LC243204

 
lab chem
LC243204
Each for $112.74

LC243205

 
lab chem
LC243205
Each for $265.57
Description & Specifications

Specifications

Formula Weight 40
Purity Grade Certified Grade
Specific Gravity 1.02g/mL
UN Number UN1824
Health Hazard 1 Danger
Synonym Caustic Soda
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 7732-18-5
Physical Form Liquid
Color Clear, colorless
CAS Max % 98.43
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 1310-73-2
Concentration 0.400N ±0.004N (0.4M)
Packaging Poly Bottle
Identification Passes Test
Linear Formula NaOH
Density 1.02g/mL
Chemical Name or Material Sodium Hydroxide
Storage Information Room Temperature
CAS Max % 1.57
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.